ENA258389671 Enamine

2-{[2,6-bis(propan-2-yl)phenyl]amino}ethan-1-ol

  • MDL number MFCD14705814
  • Cas Number : 790206-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59736
Availability details for
Quantity
  • Availability
  • Add to Cart