ENA258389683 Enamine

3-(4-methanesulfonylphenoxy)propanoic acid

  • MDL number MFCD12066109
  • Cas Number : 868277-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59764

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59764
Availability details for
Quantity
  • Availability
  • Add to Cart