ENA258389685 Enamine

2-[(pyridin-3-ylmethyl)amino]benzoic acid

  • MDL number MFCD01764307
  • Cas Number : 282719-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59767

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59767
Availability details for
Quantity
  • Availability
  • Add to Cart