ENA258389686 Enamine

4-(1,2,3,4-tetrahydroisoquinolin-2-yl)benzoic acid

  • MDL number MFCD11922635
  • Cas Number : 1020933-05-4
  • Molecular Weight : 253.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59768
Availability details for
Quantity
  • Availability
  • Add to Cart