ENA258389689 Enamine

4-{methyl[(4-methylphenyl)methyl]amino}benzoic acid

  • MDL number MFCD11923145
  • Cas Number : 1036502-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59772
Availability details for
Quantity
  • Availability
  • Add to Cart