ENA258389694 Enamine

4-(chloromethyl)-2-cyclopentyl-1,3-thiazole

  • MDL number MFCD12768695
  • Cas Number : 774158-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59781

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59781
Availability details for
Quantity
  • Availability
  • Add to Cart