ENA258389703 Enamine

octahydro-1H-pyrrolo[1,2-a][1,4]diazepin-1-one

  • MDL number MFCD09832104
  • Cas Number : 109324-81-4
  • Molecular Weight : 154.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59792
Availability details for
Quantity
  • Availability
  • Add to Cart