ENA258389706 Enamine

8-(trifluoromethoxy)quinolin-4-ol

  • MDL number MFCD03407893
  • Cas Number : 40516-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59822
Availability details for
Quantity
  • Availability
  • Add to Cart