ENA258389707 Enamine

2-[(2,5-dioxopyrrolidin-1-yl)methyl]benzonitrile

  • MDL number MFCD09927769
  • Cas Number : 1016687-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59823

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59823
Availability details for
Quantity
  • Availability
  • Add to Cart