ENA258389708 Enamine

2-(morpholin-4-yl)benzonitrile

  • MDL number MFCD01313390
  • Cas Number : 204078-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-51
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59825
Availability details for
Quantity
  • Availability
  • Add to Cart