ENA258389713 Enamine

4-(5-phenyl-2H-1,2,3,4-tetrazol-2-yl)butanoic acid

  • MDL number MFCD01648283
  • Cas Number : 21054-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59832
Availability details for
Quantity
  • Availability
  • Add to Cart