ENA258389722 Enamine

1-phenylcyclopentan-1-ol

  • MDL number MFCD00060796
  • Cas Number : 10487-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59843
Availability details for
Quantity
  • Availability
  • Add to Cart