ENA258389727 Enamine

1-(4-fluorophenyl)-3-(phenylsulfanyl)propan-1-one

  • MDL number MFCD12059003
  • Cas Number : 1157611-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59848
Availability details for
Quantity
  • Availability
  • Add to Cart