ENA258389733 Enamine

5-methyl-4-nitro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD08456975
  • Cas Number : 5334-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59856

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59856
Availability details for
Quantity
  • Availability
  • Add to Cart