ENA258389736 Enamine

1-(4-fluorophenyl)-3-(pyridin-2-ylsulfanyl)propan-1-one

  • MDL number MFCD12059031
  • Cas Number : 1157733-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59867
Availability details for
Quantity
  • Availability
  • Add to Cart