ENA258389738 Enamine

2,2,2-trifluoroethyl N-(isoquinolin-5-yl)carbamate

  • MDL number MFCD12650265
  • Cas Number : 1152953-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54221
Availability details for
Quantity
  • Availability
  • Add to Cart