ENA258389741 Enamine

2-(methanesulfonylmethyl)aniline

  • MDL number MFCD12823721
  • Cas Number : 25195-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59872

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59872
Availability details for
Quantity
  • Availability
  • Add to Cart