ENA258389743 Enamine

8-fluoroquinoline-5-sulfonamide

  • MDL number MFCD11639142
  • Cas Number : 1096846-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59876
Availability details for
Quantity
  • Availability
  • Add to Cart