ENA258389747 Enamine

4-chloro-2-cyclopropyl-5-phenylthieno[2,3-d]pyrimidine

  • MDL number MFCD12531439
  • Cas Number : 1019384-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59881

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59881
Availability details for
Quantity
  • Availability
  • Add to Cart