ENA258389748 Enamine

4-chloro-N-(piperidin-4-yl)benzene-1-sulfonamide

  • MDL number MFCD08059703
  • Cas Number : 152627-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59882
Availability details for
Quantity
  • Availability
  • Add to Cart