ENA258389750 Enamine

2-(1H-pyrazole-3-amido)benzoic acid

  • MDL number MFCD00600615
  • Cas Number : 405278-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54337
Availability details for
Quantity
  • Availability
  • Add to Cart