ENA258389752 Enamine

5,6-difluoro-1,3-benzothiazol-2-amine

  • MDL number MFCD02944727
  • Cas Number : 352214-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171...173
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59886

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-59886
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart