ENA258389753 Enamine

2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-2-phenylacetic acid

  • MDL number MFCD31813432
  • Cas Number : 777917-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54420
Availability details for
Quantity
  • Availability
  • Add to Cart