ENA258389755 Enamine

3-chloro-6-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)pyridazine

  • MDL number MFCD12078679
  • Cas Number : 1154334-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59889
Availability details for
Quantity
  • Availability
  • Add to Cart