ENA258389765 Enamine

2-[N-methyl-1-(pyridin-2-yl)formamido]acetic acid

  • MDL number MFCD09048577
  • Cas Number : 125686-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54492
Availability details for
Quantity
  • Availability
  • Add to Cart