ENA258389772 Enamine

2-(2,3-dihydro-1,4-benzodioxin-6-yl)-2-methylpropanoic acid

  • MDL number MFCD12816628
  • Cas Number : 1178765-70-2
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59916
Availability details for
Quantity
  • Availability
  • Add to Cart