ENA258389773 Enamine

2-(2-methoxyphenyl)-2-methylpropanoic acid

  • MDL number MFCD11036932
  • Cas Number : 468064-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-127
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59917
Availability details for
Quantity
  • Availability
  • Add to Cart