ENA258389778 Enamine

5,7-dimethyladamantane-1,3-diol

  • Cas Number : 10347-01-0
  • Molecular Weight : 196.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54558
Availability details for
Quantity
  • Availability
  • Add to Cart