ENA258389786 Enamine

ethyl N-(6-amino-1,3-benzothiazol-2-yl)carbamate

  • MDL number MFCD09803607
  • Cas Number : 1016526-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-59934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59934
Availability details for
Quantity
  • Availability
  • Add to Cart