ENA258389788 Enamine

2-amino-2-(3-bromophenyl)acetamide

  • MDL number MFCD12151944
  • Cas Number : 1105679-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107...109
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59936

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59936
Availability details for
Quantity
  • Availability
  • Add to Cart