ENA258389790 Enamine

potassium 3-[4-(morpholine-4-sulfonyl)phenyl]propanoate

  • MDL number MFCD08270251
  • Cas Number : 1007012-15-8
  • Molecular Weight : 337.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-21396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21396
Availability details for
Quantity
  • Availability
  • Add to Cart