ENA258389792 Enamine

4-(aminomethyl)-2-fluoro-N,N-dimethylaniline

  • MDL number MFCD09907412
  • Cas Number : 951907-17-8
  • Molecular Weight : 168.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59941

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59941
Availability details for
Quantity
  • Availability
  • Add to Cart