ENA258389795 Enamine

2-(1,3-dioxaindan-5-yl)-2-methylpropan-1-amine

  • MDL number MFCD12774756
  • Cas Number : 23131-47-7
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59945
Availability details for
Quantity
  • Availability
  • Add to Cart