ENA258389797 Enamine

N-(1-acetylpiperidin-4-yl)-2-chloroacetamide

  • MDL number MFCD12147349
  • Cas Number : 1152621-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23226
Availability details for
Quantity
  • Availability
  • Add to Cart