ENA258389804 Enamine

8-bromo-6-chloro-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD11207564
  • Cas Number : 890839-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54705
Availability details for
Quantity
  • Availability
  • Add to Cart