ENA258389806 Enamine

4-(cyanomethoxy)-5-methoxy-2-nitrobenzoic acid

  • MDL number MFCD11536189
  • Cas Number : 1096802-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54906
Availability details for
Quantity
  • Availability
  • Add to Cart