ENA258389814 Enamine

2-(3-methyl-1,2,4-oxadiazol-5-yl)ethan-1-amine hydrochloride

  • MDL number MFCD12026513
  • Cas Number : 1185295-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59970
Availability details for
Quantity
  • Availability
  • Add to Cart