ENA258389816 Enamine

thiomorpholine-4-carbohydrazide

  • MDL number MFCD11212703
  • Cas Number : 1005420-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59972
Availability details for
Quantity
  • Availability
  • Add to Cart