ENA258389818 Enamine

1-(2-chlorophenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD11168140
  • Cas Number : 1038261-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133...135
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59974

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59974
Availability details for
Quantity
  • Availability
  • Add to Cart