ENA258389819 Enamine

3-(methanesulfonylmethyl)aniline

  • MDL number MFCD12776627
  • Cas Number : 261925-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-127
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59975
Availability details for
Quantity
  • Availability
  • Add to Cart