ENA258389822 Enamine

2-(chloromethyl)pyrimidin-1-ium-1-olate

  • MDL number MFCD14705853
  • Cas Number : 206882-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59979
Availability details for
Quantity
  • Availability
  • Add to Cart