ENA258389823 Enamine

2-(trimethyl-1H-pyrazol-4-yl)propanoic acid

  • MDL number MFCD08700816
  • Cas Number : 1007460-61-8
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59983
Availability details for
Quantity
  • Availability
  • Add to Cart