ENA258389829 Enamine

(naphthalen-1-yl)methanethiol

  • MDL number MFCD00046448
  • Cas Number : 5254-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59990
Availability details for
Quantity
  • Availability
  • Add to Cart