ENA258389831 Enamine

2-(cyclohexylmethyl)pyrrolidine

  • MDL number MFCD02663459
  • Cas Number : 60601-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59992
Availability details for
Quantity
  • Availability
  • Add to Cart