ENA258389844 Enamine

N'-hydroxy-2-oxo-1,2-dihydropyridine-3-carboximidamide

  • MDL number MFCD11213717
  • Cas Number : 1241704-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176...178
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60012

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60012
Availability details for
Quantity
  • Availability
  • Add to Cart