ENA258389846 Enamine

5-bromo-1-(prop-2-yn-1-yl)-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD02108126
  • Cas Number : 79552-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60014

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60014
Availability details for
Quantity
  • Availability
  • Add to Cart