ENA258389847 Enamine

N-(6-amino-1,3-benzothiazol-2-yl)methanesulfonamide

  • MDL number MFCD09738124
  • Cas Number : 954270-87-2
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60015
Availability details for
Quantity
  • Availability
  • Add to Cart