ENA258389848 Enamine

2-(3-hydroxyphenyl)acetamide

  • MDL number MFCD06656129
  • Cas Number : 22446-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-123
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60016
Availability details for
Quantity
  • Availability
  • Add to Cart