ENA258389849 Enamine

3-[5-(4-methoxyphenyl)-1,3-oxazol-2-yl]propanoic acid

  • MDL number MFCD11107672
  • Cas Number : 1019357-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-26339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26339
Availability details for
Quantity
  • Availability
  • Add to Cart