ENA258389851 Enamine

1-(2-bromoethyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD09805337
  • Cas Number : 1016508-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60023

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60023
Availability details for
Quantity
  • Availability
  • Add to Cart